ENAH2C55725C Enamine

tert-butyl 3-benzyl-4-oxopyrrolidine-1-carboxylate

  • MDL number MFCD24470271
  • Cas Number : 897046-05-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008700

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008700
Availability details for
Quantity
  • Availability
  • Add to Cart