ENAH2C55725F Enamine

8-(hydroxymethyl)-1,3-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione

  • MDL number MFCD03258838
  • Cas Number : 2879-16-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 243-244
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008832

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008832
Availability details for
Quantity
  • Availability
  • Add to Cart