ENAH2C557263 Enamine

3-methyl-5-phenyl-1H-indole-2-carboxylic acid

  • MDL number MFCD20229959
  • Cas Number : 191868-97-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008865

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008865
Availability details for
Quantity
  • Availability
  • Add to Cart