ENAH2C557271 Enamine

(2-chloro-6-methanesulfonylphenyl)methanol

  • MDL number MFCD26007852
  • Cas Number : 1549395-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008922

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008922
Availability details for
Quantity
  • Availability
  • Add to Cart