ENAH2C557272 Enamine

1-{4-methyl-[2,2'-bipyrimidin]-5-yl}ethan-1-one

  • MDL number MFCD17216926
  • Cas Number : 1342643-17-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008923

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008923
Availability details for
Quantity
  • Availability
  • Add to Cart