ENAH2C557276 Enamine

methyl 2-amino-3-(5-methoxy-1H-indol-3-yl)propanoate

  • MDL number MFCD09756113
  • Cas Number : 176972-06-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008931

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008931
Availability details for
Quantity
  • Availability
  • Add to Cart