ENAH2C557278 Enamine

methyl 2-(4-bromophenyl)-2-diazoacetate

  • MDL number MFCD28386133
  • Cas Number : 264881-99-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008935

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008935
Availability details for
Quantity
  • Availability
  • Add to Cart