ENAH2C55727B Enamine

2-oxo-1-(2,2,2-trifluoroethyl)-1,2-dihydropyridine-4-carboxylic acid

  • MDL number MFCD24326445
  • Cas Number : 1203544-08-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008939

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008939
Availability details for
Quantity
  • Availability
  • Add to Cart