ENAH2C557283 Enamine

3-hydroxy-3,4-dihydro-2H-1-benzopyran-4-one

  • MDL number MFCD18450992
  • Cas Number : 74736-89-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008965

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008965
Availability details for
Quantity
  • Availability
  • Add to Cart