ENAH2C557286 Enamine

3-[(benzyloxy)methyl]cyclobutan-1-ol

  • MDL number MFCD12761270
  • Cas Number : 197167-53-6
  • Molecular Weight : 192.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-[(benzyloxy)methyl]cyclobutanol
Inchi Code 1S/C12H16O2/c13-12-6-11(7-12)9-14-8-10-4-2-1-3-5-10/h1-5,11-13H,6-9H2
Inchi Key KVQOEODTOJCIPA-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-2008968

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-2008968
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart