ENAH2C557287 Enamine

dimethoxy-1lambda4,2,5-thiadiazol-1-one

  • MDL number MFCD21604155
  • Cas Number : 78441-23-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-137
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008969

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008969
Availability details for
Quantity
  • Availability
  • Add to Cart