ENAH2C557288 Enamine

1-ethenyl-1H-1,2,3,4-tetrazole

  • MDL number MFCD09033071
  • Cas Number : 17578-18-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008974

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008974
Availability details for
Quantity
  • Availability
  • Add to Cart