ENAH2C55728F Enamine

3,4-dihydro-1H-2-benzopyran-7-ol

  • MDL number MFCD22536232
  • Cas Number : 1391209-22-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008987

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008987
Availability details for
Quantity
  • Availability
  • Add to Cart