ENAH2C557293 Enamine

5-oxo-2-phenyl-1-propylpyrrolidine-3-carboxylic acid

  • MDL number MFCD15730825
  • Cas Number : 1267035-97-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009003

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009003
Availability details for
Quantity
  • Availability
  • Add to Cart