ENAH2C557294 Enamine

N-(2-hydroxyethyl)cyclopropanesulfonamide

  • MDL number MFCD20352463
  • Cas Number : 1534388-85-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009004

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009004
Availability details for
Quantity
  • Availability
  • Add to Cart