ENAH2C557295 Enamine

1-methyl-2,5-dihydro-1H-pyrrole

  • MDL number MFCD16038583
  • Cas Number : 554-15-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009005

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009005
Availability details for
Quantity
  • Availability
  • Add to Cart