ENAH2C557296 Enamine

methyl 3-chloro-2-methyl-3-oxopropanoate

  • MDL number MFCD28655969
  • Cas Number : 23185-24-2
  • Molecular Weight : 150.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 3-chloro-2-methyl-3-oxopropanoate
Inchi Code 1S/C5H7ClO3/c1-3(4(6)7)5(8)9-2/h3H,1-2H3
Inchi Key OWRCJIZVGNZKDU-UHFFFAOYSA-N

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-2009006

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-2009006
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart