ENAH2C557299 Enamine

1-methyl-3-(piperidin-3-yl)-1H-1,2,4-triazol-5-amine dihydrochloride

  • MDL number MFCD31667185
  • Cas Number : 2230807-04-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009018

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009018
Availability details for
Quantity
  • Availability
  • Add to Cart