ENAH2C5572A0 Enamine

2-(aminooxy)propanenitrile

  • MDL number MFCD21921021
  • Cas Number : 1510417-69-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009033

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009033
Availability details for
Quantity
  • Availability
  • Add to Cart