ENAH2C5572B2 Enamine

[1-methyl-5-(trifluoromethyl)-1H-pyrazol-4-yl]boronic acid

  • MDL number MFCD30188652
  • Cas Number : 2246648-49-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009091

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009091
Availability details for
Quantity
  • Availability
  • Add to Cart