ENAH2C5572B9 Enamine

8-bromo-7-chloro-1,2,3,4-tetrahydroisoquinoline hydrochloride

  • MDL number MFCD31667190
  • Cas Number : 2230807-08-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009099

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009099
Availability details for
Quantity
  • Availability
  • Add to Cart