ENAH2C5572BB Enamine

1-[(3-bromophenyl)methyl]piperidin-4-ol

  • MDL number MFCD00977077
  • Cas Number : 184921-06-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009102

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009102
Availability details for
Quantity
  • Availability
  • Add to Cart