ENAH2C5572BC Enamine

3-[3-(trifluoromethyl)phenyl]pyrrolidin-3-ol hydrochloride

  • MDL number MFCD01687419
  • Cas Number : 1744-48-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009103

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009103
Availability details for
Quantity
  • Availability
  • Add to Cart