ENAH2C5572C0 Enamine

(6-chloronaphthalen-2-yl)boronic acid

  • MDL number MFCD18411299
  • Cas Number : 870822-86-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009110

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009110
Availability details for
Quantity
  • Availability
  • Add to Cart