ENAH2C5572C1 Enamine

(5-amino-1-methyl-1H-pyrazol-3-yl)methanol

  • MDL number MFCD25949426
  • Cas Number : 1224888-30-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009111

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009111
Availability details for
Quantity
  • Availability
  • Add to Cart