ENAH2C5572C3 Enamine

methyl 4-bromo-3-methyl-1H-pyrrole-2-carboxylate

  • MDL number MFCD11557220
  • Cas Number : 1092286-09-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009118

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009118
Availability details for
Quantity
  • Availability
  • Add to Cart