ENAH2C5572C6 Enamine

5-amino-2-(1-{[(tert-butoxy)carbonyl]amino}ethyl)-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD31667192
  • Cas Number : 2230799-73-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009123

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009123
Availability details for
Quantity
  • Availability
  • Add to Cart