ENAH2C5572C9 Enamine

tert-butyl 8-(aminomethyl)-6-azabicyclo[3.2.1]octane-6-carboxylate hydrochloride

  • MDL number MFCD31667194
  • Cas Number : 2230798-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009130

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009130
Availability details for
Quantity
  • Availability
  • Add to Cart