ENAH2C5572CE Enamine

1-[(4-fluoro-3-methylphenyl)methyl]cyclopropan-1-amine hydrochloride

  • MDL number MFCD31667196
  • Cas Number : 2230807-47-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009139

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009139
Availability details for
Quantity
  • Availability
  • Add to Cart