ENAH2C5572D4 Enamine

2-(5,5-dimethyl-3-oxo-1,2-oxazolidin-2-yl)acetic acid

  • MDL number MFCD31667848
  • Cas Number : 1784436-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009148

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009148
Availability details for
Quantity
  • Availability
  • Add to Cart