ENAH2C5572D9 Enamine

(4R)-6-(propan-2-yl)-3,4-dihydro-2H-1-benzopyran-4-ol

  • MDL number MFCD21295414
  • Cas Number : 1567968-24-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009168

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009168
Availability details for
Quantity
  • Availability
  • Add to Cart