ENAH2C5572DE Enamine

2-bromo-3-(bromomethyl)-1-fluoro-4-nitrobenzene

  • MDL number MFCD28740168
  • Cas Number : 1804843-96-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009190

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009190
Availability details for
Quantity
  • Availability
  • Add to Cart