ENAH2C5572E8 Enamine

4-(4,5,6,7-tetrahydro-1H-indazol-3-yl)aniline dihydrochloride

  • MDL number MFCD31667245
  • Cas Number : 2230807-93-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009252

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009252
Availability details for
Quantity
  • Availability
  • Add to Cart