ENAH2C5572EA Enamine

3-(piperazin-1-yl)pentanenitrile dihydrochloride

  • MDL number MFCD31667849
  • Cas Number : 2230807-40-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009260

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009260
Availability details for
Quantity
  • Availability
  • Add to Cart