ENAH2C5572F0 Enamine

3-(2-hydroxyethoxy)-4-methoxybenzaldehyde

  • MDL number MFCD08696459
  • Cas Number : 57643-36-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009274

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009274
Availability details for
Quantity
  • Availability
  • Add to Cart