ENAH2C5572F3 Enamine

methyl 3-oxobicyclo[3.2.1]octane-2-carboxylate

  • MDL number MFCD25961300
  • Cas Number : 228117-37-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009278

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009278
Availability details for
Quantity
  • Availability
  • Add to Cart