ENAH2C5572F6 Enamine

(1r,3r)-3-[(1H-pyrazol-1-yl)methyl]cyclobutan-1-amine dihydrochloride

  • MDL number MFCD31667248
  • Cas Number : 2230789-59-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009282

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009282
Availability details for
Quantity
  • Availability
  • Add to Cart