ENAH2C5572FE Enamine

3,4-dimethoxythiophene-2-carboxylic acid

  • MDL number MFCD20645208
  • Cas Number : 113589-62-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009291

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009291
Availability details for
Quantity
  • Availability
  • Add to Cart