ENAH2C557302 Enamine

tert-butyl 3-(3-hydroxyazetidin-1-yl)pyrrolidine-1-carboxylate

  • MDL number MFCD28619665
  • Cas Number : 1781982-51-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009300

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009300
Availability details for
Quantity
  • Availability
  • Add to Cart