ENAH2C557306 Enamine

4-[(prop-2-en-1-yl)carbamoyl]benzoic acid

  • MDL number MFCD12173470
  • Cas Number : 1085698-16-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009330

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009330
Availability details for
Quantity
  • Availability
  • Add to Cart