ENAH2C557310 Enamine

(4R)-6-(propan-2-yl)-3,4-dihydro-2H-1-benzopyran-4-amine hydrochloride

  • MDL number MFCD22392660
  • Cas Number : 676134-24-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009379

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009379
Availability details for
Quantity
  • Availability
  • Add to Cart