ENAH2C557317 Enamine

[5-bromo-3-(hydroxymethyl)pyridin-2-yl]methanol hydrochloride

  • MDL number MFCD20527344
  • Cas Number : 1356108-96-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009398

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009398
Availability details for
Quantity
  • Availability
  • Add to Cart