ENAH2C557318 Enamine

2-[5-(aminomethyl)-1H-1,2,4-triazol-3-yl]propan-2-ol hydrochloride

  • MDL number MFCD31667252
  • Cas Number : 2230798-90-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009402

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009402
Availability details for
Quantity
  • Availability
  • Add to Cart