ENAH2C55731C Enamine

(5-chloro-2,3-dihydro-1-benzofuran-7-yl)(cyclopropyl)methanol

  • MDL number MFCD26904524
  • Cas Number : 1566939-13-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1721173

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1721173
Availability details for
Quantity
  • Availability
  • Add to Cart