ENAH2C55731D Enamine

2-[(4-bromophenyl)methyl]-6-methoxyhexanoic acid

  • MDL number MFCD26329744
  • Cas Number : 1917420-50-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009435

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009435
Availability details for
Quantity
  • Availability
  • Add to Cart