ENAH2C55731E Enamine

2-chloro-5-nitro-6-(trifluoromethyl)pyrimidin-4-amine

  • MDL number MFCD16615889
  • Cas Number : 23663-18-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1721208

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1721208
Availability details for
Quantity
  • Availability
  • Add to Cart