ENAH2C557325 Enamine

6-cyano-5-methylpyridine-3-carboxylic acid

  • MDL number MFCD28780620
  • Cas Number : 1360953-72-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009486

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009486
Availability details for
Quantity
  • Availability
  • Add to Cart