ENAH2C557328 Enamine

5-(hydroxymethyl)-1,4-dioxan-2-one

  • MDL number MFCD28654551
  • Cas Number : 122150-99-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009496

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009496
Availability details for
Quantity
  • Availability
  • Add to Cart