ENAH2C55732B Enamine

6-chloro-1-methylisoquinoline

  • MDL number MFCD31667935
  • Cas Number : 2229934-60-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009505

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009505
Availability details for
Quantity
  • Availability
  • Add to Cart