ENAH2C557338 Enamine

[2-(tert-butoxy)ethyl](ethyl)amine

  • MDL number MFCD16076420
  • Cas Number : 1248963-53-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1721395

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1721395
Availability details for
Quantity
  • Availability
  • Add to Cart